Learn More
Biomatik Corporation Recombinant Lymantria dispar multicapsid nuclear polyhedrosis virus Major capsid protein (P39), 100UG
Supplier:  Biomatik Corporation RPC22512
Encompass_Preferred
Recombinant Lymantria dispar multicapsid nuclear polyhedrosis virus Major capsid protein (P39) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Lymantria dispar multicapsid nuclear polyhedrosis virus (LdMNPV). Target Name: P39. Target Synonyms: P39; Major capsid protein. Accession Number: P35840. Expression Region: 1~356aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 44.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5136