Learn More
Biomatik Corporation Recombinant Glycine max Ribulose bisphosphate carboxylase large chain (rbcL), 100UG
Supplier:  Biomatik Corporation RPC22519
Encompass_Preferred
Recombinant Glycine max Ribulose bisphosphate carboxylase large chain (rbcL) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Glycine max (Soybean) (Glycine hispida). Target Name: rbcL. Target Synonyms: RuBisCO large subunit. Accession Number: P27066. Expression Region: 3~475aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 68.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5137