Learn More
Biomatik Corporation Recombinant Human ATP synthase subunit alpha, mitochondrial (ATP5F1A), 100UG
Supplier:  Biomatik Corporation RPC20007
Encompass_Preferred
Recombinant Human ATP synthase subunit alpha, mitochondrial (ATP5F1A) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ATP5F1A. Target Synonyms: ATP synthase alpha chain; ATP synthase alpha chain; mitochondrial; ATP synthase subunit alpha; ATP synthase subunit alpha mitochondrial; ATP synthase; H+ transporting; mitochondrial F1 complex; alpha subunit 1; cardiac muscle. Accession Number: P25705. Expression Region: 44~553aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 59.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4536