Learn More
Biomatik Corporation Recombinant Crotalus adamanteus Snake venom metalloproteinase adamalysin-2, 100UG
Supplier:  Biomatik Corporation RPC22529
Encompass_Preferred
Recombinant Crotalus adamanteus Snake venom metalloproteinase adamalysin-2 is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Crotalus adamanteus (Eastern diamondback rattlesnake). Target Name: Crotalus adamanteus Snake venom metalloproteinase adamalysin-2. Target Synonyms: Adamalysin IIProteinase II. Accession Number: P34179. Expression Region: 1~203aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 27.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5140