Learn More
Biomatik Corporation Recombinant Salmonella typhimurium Protein PrgJ (PrgJ), 100UG
Catalog No. 501505141
Encompass_Preferred
Recombinant Salmonella typhimurium Protein PrgJ (PrgJ) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Salmonella typhimurium (strain LT2 / SGSC1412 / ATCC 700720). Target Name: prgJ. Target Synonyms: prgJ; STM2872Protein PrgJ. Accession Number: P41785. Expression Region: 1~101aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 15.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5141 Supplier Biomatik Corporation Supplier No. RPC22532
Recombinant Salmonella typhimurium Protein PrgJ (PrgJ) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Salmonella typhimurium (strain LT2 / SGSC1412 / ATCC 700720). Target Name: prgJ. Target Synonyms: prgJ; STM2872Protein PrgJ. Accession Number: P41785. Expression Region: 1~101aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 15.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.