Learn More
Biomatik Corporation Recombinant Rabies virus Matrix protein (M), 100UG
Catalog No. 501505143
Encompass_Preferred
Recombinant Rabies virus Matrix protein (M) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rabies virus (strain CVS-11) (RABV). Target Name: M. Target Synonyms: Phosphoprotein M2. Accession Number: P25223. Expression Region: 1~202aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 28.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5143 Supplier Biomatik Corporation Supplier No. RPC22543
Recombinant Rabies virus Matrix protein (M) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Rabies virus (strain CVS-11) (RABV). Target Name: M. Target Synonyms: Phosphoprotein M2. Accession Number: P25223. Expression Region: 1~202aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 28.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.