Learn More
Biomatik Corporation Recombinant Actinoplanes utahensis Aculeacin-A acylase (aac), partial, 100UG
Supplier:  Biomatik Corporation RPC22549
Encompass_Preferred
Recombinant Actinoplanes utahensis Aculeacin-A acylase (aac), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Actinoplanes utahensis. Target Name: aac. Target Synonyms: Aculeacin-A acylase small subunitAculeacin-A acylase large subunit. Accession Number: P29958. Expression Region: 35~214aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 35.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5144