Learn More
Biomatik Corporation Recombinant Newcastle disease virus Hemagglutinin-neuraminidase (HN), partial, 100UG
Supplier:  Biomatik Corporation RPC22571
Encompass_Preferred
Recombinant Newcastle disease virus Hemagglutinin-neuraminidase (HN), partial is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Newcastle disease virus (strain Her/33) (NDV). Target Name: HN. Target Synonyms: HN; Hemagglutinin-neuraminidase; EC 3.2.1.18. Accession Number: P35741. Expression Region: 115~473aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 44.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5147