Learn More
Biomatik Corporation Recombinant Macaca fascicularis Transthyretin (TTR), 100UG
Supplier:  Biomatik Corporation RPC20100
Encompass_Preferred
Recombinant Macaca fascicularis Transthyretin (TTR) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Macaca fascicularis (Crab-eating macaque) (Cynomolgus monkey). Target Name: TTR. Target Synonyms: Prealbumin. Accession Number: Q8HXW1. Expression Region: 21~147aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 17.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4591