Learn More
Biomatik Corporation Recombinant Betula pendula Calcium-binding allergen Bet v 3 (BETVIII), 100UG
Supplier:  Biomatik Corporation RPC22577
Encompass_Preferred
Recombinant Betula pendula Calcium-binding allergen Bet v 3 (BETVIII) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Betula pendula (European white birch)(Betula verrucosa). Target Name: BETVIII. Target Synonyms: Allergen Bet v IIIAllergen: Bet v 3. Accession Number: P43187. Expression Region: 1~205aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 43.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5150