Learn More
Biomatik Corporation Recombinant Yersinia pestis F1 capsule antigen (caf1), 100UG
Supplier:  Biomatik Corporation RPC22584
Encompass_Preferred
Recombinant Yersinia pestis F1 capsule antigen (caf1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Yersinia pestis. Target Name: caf1. Target Synonyms: caf1; YPMT1.84; Y1100; YP_pMT082F1 capsule antigen. Accession Number: P26948. Expression Region: 22~170aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 19.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5153