Learn More
Biomatik Corporation Recombinant Staphylococcus aureus Enterotoxin type C-2 (entC2), 100UG
Supplier:  Biomatik Corporation RPC22590
Encompass_Preferred
Recombinant Staphylococcus aureus Enterotoxin type C-2 (entC2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Staphylococcus aureus. Target Name: entC2. Target Synonyms: SEC2. Accession Number: P34071. Expression Region: 28~266aa. Tag Info: Tag-Free. Theoretical MW: 27.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5156