Learn More
Biomatik Corporation Recombinant Bovine Cathelicidin-4 (CATHL4), 100UG
Supplier:  Biomatik Corporation RPC22605
Encompass_Preferred
Recombinant Bovine Cathelicidin-4 (CATHL4) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bovine (Bos taurus). Target Name: CATHL4. Target Synonyms: Indolicidin. Accession Number: P33046. Expression Region: 131~143aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 17.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5157