Learn More
Biomatik Corporation Recombinant Cricetulus griseus Actin, cytoplasmic 1 (ACTB), 100UG
Supplier:  Biomatik Corporation RPC22611
Encompass_Preferred
Recombinant Cricetulus griseus Actin, cytoplasmic 1 (ACTB) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Cricetulus griseus (Chinese hamster) (Cricetulus barabensis griseus). Target Name: ACTB. Target Synonyms: Beta-actin. Accession Number: P48975. Expression Region: 1~375aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 45.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5159