Learn More
Biomatik Corporation Recombinant Chicken anemia virus Apoptin (VP3), 100UG
Supplier:  Biomatik Corporation RPC22625
Encompass_Preferred
Recombinant Chicken anemia virus Apoptin (VP3) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Chicken anemia virus (isolate USA 26p4) (CAV). Target Name: VP3. Target Synonyms: VP3; Apoptin. Accession Number: P54095. Expression Region: 1~120aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 17.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5161