Learn More
Biomatik Corporation Recombinant Mannheimia haemolytica Leukotoxin (lktA), partial, 100UG
Supplier:  Biomatik Corporation RPC22627
Encompass_Preferred
Recombinant Mannheimia haemolytica Leukotoxin (lktA), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mannheimia haemolytica (Pasteurella haemolytica). Target Name: lktA. Target Synonyms: lktALeukotoxin; Lkt. Accession Number: P55118. Expression Region: 1~229aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 29.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5162