Learn More
Biomatik Corporation Recombinant Human Heterogeneous nuclear ribonucleoprotein M (HNRNPM), 100UG
Catalog No. 501504593
Encompass_Preferred
Recombinant Human Heterogeneous nuclear ribonucleoprotein M (HNRNPM) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HNRNPM. Target Synonyms: CEA receptor; CEAR; Heterogeneous nuclear ribonucleoprotein M; Heterogenous nuclear ribonucleoprotein M4; hnRNA binding protein M4; hnRNP M; Hnrnpm; HNRNPM4; HNRPM; HNRPM_HUMAN; HNRPM4; HTGR1; M3; M4; M4 protein; N-acetylglucosamine receptor 1; NAGR1. Accession Number: P52272. Expression Region: 2~730aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 93.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4593 Supplier Biomatik Corporation Supplier No. RPC20103
Recombinant Human Heterogeneous nuclear ribonucleoprotein M (HNRNPM) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HNRNPM. Target Synonyms: CEA receptor; CEAR; Heterogeneous nuclear ribonucleoprotein M; Heterogenous nuclear ribonucleoprotein M4; hnRNA binding protein M4; hnRNP M; Hnrnpm; HNRNPM4; HNRPM; HNRPM_HUMAN; HNRPM4; HTGR1; M3; M4; M4 protein; N-acetylglucosamine receptor 1; NAGR1. Accession Number: P52272. Expression Region: 2~730aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 93.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.