Learn More
Biomatik Corporation Recombinant Enterobacteria phage 933W Shiga-like toxin 2 subunit B (stxB2), 100UG
Catalog No. 501505172
Encompass_Preferred
Recombinant Enterobacteria phage 933W Shiga-like toxin 2 subunit B (stxB2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Enterobacteria phage 933W (Bacteriophage 933W). Target Name: stxB2. Target Synonyms: Verocytotoxin 2 subunit B; Verotoxin 2 subunit B. Accession Number: P09386. Expression Region: 20~89aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 23.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5172 Supplier Biomatik Corporation Supplier No. RPC22708
Recombinant Enterobacteria phage 933W Shiga-like toxin 2 subunit B (stxB2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Enterobacteria phage 933W (Bacteriophage 933W). Target Name: stxB2. Target Synonyms: Verocytotoxin 2 subunit B; Verotoxin 2 subunit B. Accession Number: P09386. Expression Region: 20~89aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 23.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.