Learn More
Biomatik Corporation Recombinant Trichosanthes kirilowii Ribosome-inactivating protein alpha-trichosanthin, 100UG
Supplier:  Biomatik Corporation RPC22709
Encompass_Preferred
Recombinant Trichosanthes kirilowii Ribosome-inactivating protein alpha-trichosanthin is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Trichosanthes kirilowii (Chinese snake gourd) (Chinese cucumber). Target Name: Trichosanthes kirilowii Ribosome-inactivating protein alpha-trichosanthin. Target Synonyms: Ribosome-inactivating protein alpha-trichosanthin; Alpha-TCS; EC 3.2.2.22; rRNA N-glycosidase. Accession Number: P09989. Expression Region: 24~270aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 43.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5173