Learn More
Biomatik Corporation Recombinant Ustilago sphaerogena Ribonuclease U2 (RNU2), 100UG
Supplier:  Biomatik Corporation RPC22739
Encompass_Preferred
Recombinant Ustilago sphaerogena Ribonuclease U2 (RNU2) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Ustilago sphaerogena (Smut fungus). Target Name: RNU2. Target Synonyms: RNU2; Ribonuclease U2; RNase U2; EC 4.6.1.20. Accession Number: P00654. Expression Region: 1~114aa. Tag Info: N-Terminal 6Xhis-B2M-Tagged. Theoretical MW: 26.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5178