Learn More
Biomatik Corporation Recombinant Staphylococcus aureus Enterotoxin type B (entB), 100UG
Catalog No. 501505180
Encompass_Preferred
Recombinant Staphylococcus aureus Enterotoxin type B (entB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Staphylococcus aureus. Target Name: entB. Target Synonyms: SEB. Accession Number: P01552. Expression Region: 28~266aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 55.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5180 Supplier Biomatik Corporation Supplier No. RPC22746
Recombinant Staphylococcus aureus Enterotoxin type B (entB) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Staphylococcus aureus. Target Name: entB. Target Synonyms: SEB. Accession Number: P01552. Expression Region: 28~266aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 55.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.