Learn More
Biomatik Corporation Recombinant BK polyomavirus Major capsid protein VP1, 100UG
Supplier:  Biomatik Corporation RPC22755
Encompass_Preferred
Recombinant BK polyomavirus Major capsid protein VP1 is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: BK polyomavirus (BKPyV). Target Name: BK polyomavirus Major capsid protein VP1. Target Synonyms: Major structural protein VP1. Accession Number: P03088. Expression Region: 1~362aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 60.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5182