Learn More
Biomatik Corporation Recombinant Human adenovirus C serotype 5 Protease (L3), 100UG
Supplier:  Biomatik Corporation RPC22758
Encompass_Preferred
Recombinant Human adenovirus C serotype 5 Protease (L3) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human adenovirus C serotype 5 (HAdV-5) (Human adenovirus 5). Target Name: L3. Target Synonyms: AdenainAdenovirus protease. Accession Number: P03253. Expression Region: 1~204aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 28.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5184