Learn More
Biomatik Corporation Recombinant Human rhinovirus A serotype 89 Genome polyprotein, partial, 100UG
Supplier:  Biomatik Corporation RPC22779
Encompass_Preferred
Recombinant Human rhinovirus A serotype 89 Genome polyprotein is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human rhinovirus A serotype 89 (strain 41467-Gallo) (HRV-89). Target Name: Human rhinovirus A serotype 89 Genome polyprotein. Target Synonyms: Genome polyprotein; Cleaved into: P1; Capsid protein VP0; VP4-VP2; Capsid protein VP4; P1A; Virion protein 4; Capsid protein VP2. Accession Number: P07210. Expression Region: 575~866aa. Tag Info: N-Terminal 10Xhis-Tagged. Theoretical MW: 38.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5188