Learn More
Biomatik Corporation Recombinant Plasmodium falciparum Multidrug resistance protein (MDR1), partial, 100UG
Supplier:  Biomatik Corporation RPC20107
Encompass_Preferred
Recombinant Plasmodium falciparum Multidrug resistance protein (MDR1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Plasmodium falciparum (isolate FC27 / Papua New Guinea). Target Name: MDR1. Target Synonyms: Chloroquine resistance protein. Accession Number: P13568. Expression Region: 1084~1419aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 54.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4595