Learn More
Biomatik Corporation Recombinant Bacillus licheniformis Subtilisin Carlsberg (apr), 100UG
Catalog No. 501505192
Encompass_Preferred
Recombinant Bacillus licheniformis Subtilisin Carlsberg (apr) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bacillus licheniformis. Target Name: apr. Target Synonyms: subC; apr; Subtilisin Carlsberg; EC 3.4.21.62. Accession Number: P00780. Expression Region: 106~379aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged. Theoretical MW: 42.0kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5192 Supplier Biomatik Corporation Supplier No. RPC22807
Recombinant Bacillus licheniformis Subtilisin Carlsberg (apr) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bacillus licheniformis. Target Name: apr. Target Synonyms: subC; apr; Subtilisin Carlsberg; EC 3.4.21.62. Accession Number: P00780. Expression Region: 106~379aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged. Theoretical MW: 42.0kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.