Learn More
Biomatik Corporation Recombinant Hevea brasiliensis Pro-hevein (HEV1), 100UG
Catalog No. 501505193
Encompass_Preferred
Recombinant Hevea brasiliensis Pro-hevein (HEV1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Hevea brasiliensis (Para rubber tree) (Siphonia brasiliensis). Target Name: HEV1. Target Synonyms: Major hevein. Accession Number: P02877. Expression Region: 18~204aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 24.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5193 Supplier Biomatik Corporation Supplier No. RPC22815
Recombinant Hevea brasiliensis Pro-hevein (HEV1) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Hevea brasiliensis (Para rubber tree) (Siphonia brasiliensis). Target Name: HEV1. Target Synonyms: Major hevein. Accession Number: P02877. Expression Region: 18~204aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 24.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.