Learn More
Biomatik Corporation Recombinant Cenarchaeum symbiosum DNA polymerase II large subunit (polC), partial, 100UG
Catalog No. 501505197
Encompass_Preferred
Recombinant Cenarchaeum symbiosum DNA polymerase II large subunit (polC), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Cenarchaeum symbiosum (strain A). Target Name: polC. Target Synonyms: polC; CENSYa_1827DNA polymerase II large subunit; Pol II; EC 2.7.7.7; Exodeoxyribonuclease large subunit; EC 3.1.11.1. Accession Number: A0RYM0. Expression Region: 287~832aa. Tag Info: Tag-Free. Theoretical MW: 59kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5197 Supplier Biomatik Corporation Supplier No. RPC22828
Recombinant Cenarchaeum symbiosum DNA polymerase II large subunit (polC), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Cenarchaeum symbiosum (strain A). Target Name: polC. Target Synonyms: polC; CENSYa_1827DNA polymerase II large subunit; Pol II; EC 2.7.7.7; Exodeoxyribonuclease large subunit; EC 3.1.11.1. Accession Number: A0RYM0. Expression Region: 287~832aa. Tag Info: Tag-Free. Theoretical MW: 59kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.