Learn More
Biomatik Corporation Recombinant Acinetobacter baumannii Outer membrane protein Omp38 (omp38), 100UG
Supplier:  Biomatik Corporation RPC22834
Encompass_Preferred
Recombinant Acinetobacter baumannii Outer membrane protein Omp38 (omp38) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Acinetobacter baumannii (strain ATCC 17978 / CIP 53.77 / LMG 1025 / NCDC KC755 / 5377). Target Name: omp38. Target Synonyms: omp38; A1S_2840; Outer membrane protein Omp38. Accession Number: A3M8K2. Expression Region: 20~356aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 52.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5198