Learn More
Biomatik Corporation RecombinantSchistosomamansoniMajoreggantigen, 100UG
Supplier:  Biomatik Corporation RPC20108
Encompass_Preferred
Recombinant Schistosoma mansoni Major egg antigen is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Schistosoma mansoni (Blood fluke). Target Name: Schistosoma mansoni Major egg antigen. Target Synonyms: p40. Accession Number: P12812. Expression Region: 1~354aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 43.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4596