Learn More
Biomatik Corporation Recombinant Oxyuranus microlepidotus Toxin 3FTx-Oxy6, 100UG
Catalog No. 501505199
Encompass_Preferred
Recombinant Oxyuranus microlepidotus Toxin 3FTx-Oxy6 is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Oxyuranus microlepidotus (Inland taipan). Target Name: Oxyuranus microlepidotus Toxin 3FTx-Oxy6. Target Synonyms: Three-finger toxin 3FTx-Oxy6. Accession Number: A7X4T2. Expression Region: 22~81aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 26.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5199 Supplier Biomatik Corporation Supplier No. RPC22847
Recombinant Oxyuranus microlepidotus Toxin 3FTx-Oxy6 is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Oxyuranus microlepidotus (Inland taipan). Target Name: Oxyuranus microlepidotus Toxin 3FTx-Oxy6. Target Synonyms: Three-finger toxin 3FTx-Oxy6. Accession Number: A7X4T2. Expression Region: 22~81aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 26.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.