Learn More
Biomatik Corporation Recombinant Chironex fleckeri Toxin CfTX-1, 100UG
Supplier:  Biomatik Corporation RPC22848
Encompass_Preferred
Recombinant Chironex fleckeri Toxin CfTX-1 is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Chironex fleckeri (Box jellyfish). Target Name: Chironex fleckeri Toxin CfTX-1. Target Synonyms: Toxin CfTX-1; Toxin 1. Accession Number: A7L035. Expression Region: 21~456aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 65.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5200