Learn More
Biomatik Corporation Recombinant Escherichia coli O9:H4 L-lactate dehydrogenase (lldD), 100UG
Supplier:  Biomatik Corporation RPC22853
Encompass_Preferred
Recombinant Escherichia coli O9:H4 L-lactate dehydrogenase (lldD) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli O9:H4 (strain HS). Target Name: lldD. Target Synonyms: lldD; EcHS_A3817L-lactate dehydrogenase; EC 1.1. Accession Number: A8A670. Expression Region: 1~396aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 46.7kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5202