Learn More
Biomatik Corporation Recombinant Porphyromonas gingivalis Lys-gingipain (kgp), partial, 100UG
Catalog No. 501505203
Encompass_Preferred
Recombinant Porphyromonas gingivalis Lys-gingipain (kgp), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Porphyromonas gingivalis (strain ATCC 33277 / DSM 20709 / JCM 12257). Target Name: kgp. Target Synonyms: Lysine-specific cysteine proteinase Kgp. Accession Number: B2RLK2. Expression Region: 229~594aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 56.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5203 Supplier Biomatik Corporation Supplier No. RPC22859
Recombinant Porphyromonas gingivalis Lys-gingipain (kgp), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Porphyromonas gingivalis (strain ATCC 33277 / DSM 20709 / JCM 12257). Target Name: kgp. Target Synonyms: Lysine-specific cysteine proteinase Kgp. Accession Number: B2RLK2. Expression Region: 229~594aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 56.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.