Learn More
Biomatik Corporation Recombinant Laribacter hongkongensis Orotate phosphoribosyltransferase (pyrE), 100UG
Catalog No. 501505204
Encompass_Preferred
Recombinant Laribacter hongkongensis Orotate phosphoribosyltransferase (pyrE) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Laribacter hongkongensis (strain HLHK9). Target Name: pyrE. Target Synonyms: OPRTOPRTase. Accession Number: C1D6F5. Expression Region: 1~213aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 38.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5204 Supplier Biomatik Corporation Supplier No. RPC22866
Recombinant Laribacter hongkongensis Orotate phosphoribosyltransferase (pyrE) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Laribacter hongkongensis (strain HLHK9). Target Name: pyrE. Target Synonyms: OPRTOPRTase. Accession Number: C1D6F5. Expression Region: 1~213aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 38.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.