Learn More
Biomatik Corporation Recombinant Escherichia coli Lipoate-protein ligase A (lplA), 100UG
Catalog No. 501505205
Encompass_Preferred
Recombinant Escherichia coli Lipoate-protein ligase A (lplA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12 / MC4100 / BW2952). Target Name: lplA. Target Synonyms: Lipoate--protein ligase. Accession Number: C4ZT68. Expression Region: 1~338aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 53.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5205 Supplier Biomatik Corporation Supplier No. RPC22870
Recombinant Escherichia coli Lipoate-protein ligase A (lplA) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12 / MC4100 / BW2952). Target Name: lplA. Target Synonyms: Lipoate--protein ligase. Accession Number: C4ZT68. Expression Region: 1~338aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 53.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.