Learn More
Biomatik Corporation Recombinant Mycobacterium tuberculosis Hypoxic response protein 1 (hrp1), 100UG
Supplier:  Biomatik Corporation RPC22877
Encompass_Preferred
Recombinant Mycobacterium tuberculosis Hypoxic response protein 1 (hrp1) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mycobacterium tuberculosis (strain ATCC 25618 / H37Rv). Target Name: hrp1. Target Synonyms: hrp1; Rv2626cHypoxic response protein 1; HRP1. Accession Number: P9WJA3. Expression Region: 1~143aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 35.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5206