Learn More
Biomatik Corporation Recombinant Mycobacterium tuberculosis Mycothione reductase (mtr), 100UG
Catalog No. 501505207
Encompass_Preferred
Recombinant Mycobacterium tuberculosis Mycothione reductase (mtr) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh). Target Name: mtr. Target Synonyms: Mycothiol-disulfide reductaseNADPH-dependent mycothione reductase. Accession Number: P9WHH2. Expression Region: 1~459aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 69.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5207 Supplier Biomatik Corporation Supplier No. RPC22878
Recombinant Mycobacterium tuberculosis Mycothione reductase (mtr) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mycobacterium tuberculosis (strain CDC 1551 / Oshkosh). Target Name: mtr. Target Synonyms: Mycothiol-disulfide reductaseNADPH-dependent mycothione reductase. Accession Number: P9WHH2. Expression Region: 1~459aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 69.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.