Learn More
Biomatik Corporation Recombinant Human cytomegalovirus Viral interleukin-10 homolog (UL111A), 100UG
Supplier:  Biomatik Corporation RPC22881
Encompass_Preferred
Recombinant Human cytomegalovirus Viral interleukin-10 homolog (UL111A) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human cytomegalovirus (strain Merlin) (HHV-5) (Human herpesvirus 5). Target Name: UL111A. Target Synonyms: UL111A; Viral interleukin-10 homolog; cmvIL-10; vIL-10. Accession Number: F5HC71. Expression Region: 26~176aa. Tag Info: Tag-Free. Theoretical MW: 17.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5208