Learn More
Biomatik Corporation Recombinant Human ZW10 interactor (ZWINT), 100UG
Supplier:  Biomatik Corporation RPC22917
Encompass_Preferred
Recombinant Human ZW10 interactor (ZWINT) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ZWINT. Target Synonyms: ZW10-interacting protein 1. Accession Number: O95229. Expression Region: 1~277aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 58.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5213