Learn More
Biomatik Corporation Recombinant Taraxacum officinale Root allergen protein, 100UG
Catalog No. 501505214
Encompass_Preferred
Recombinant Taraxacum officinale Root allergen protein is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Taraxacum officinale (Common dandelion) (Leontodon taraxacum). Target Name: Taraxacum officinale Root allergen protein. Target Synonyms: RAP. Accession Number: O49065. Expression Region: 1~157aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 37kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5214 Supplier Biomatik Corporation Supplier No. RPC22923
Recombinant Taraxacum officinale Root allergen protein is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Taraxacum officinale (Common dandelion) (Leontodon taraxacum). Target Name: Taraxacum officinale Root allergen protein. Target Synonyms: RAP. Accession Number: O49065. Expression Region: 1~157aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 37kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.