Learn More
Biomatik Corporation Recombinant Escherichia coli O6:K15:H31 Cysteine desulfurase IscS (iscS), 100UG
Supplier:  Biomatik Corporation RPC22957
Encompass_Preferred
Recombinant Escherichia coli O6:K15:H31 Cysteine desulfurase IscS (iscS) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli O6:K15:H31 (strain 536 / UPEC). Target Name: ISCS. Target Synonyms: iscS; ECP_2535; Cysteine desulfurase IscS; EC 2.8.1.7. Accession Number: Q0TEV5. Expression Region: 1~404aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 65.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5216