Learn More
Biomatik Corporation Recombinant Human CD166 antigen (ALCAM), partial, 100UG
Supplier:  Biomatik Corporation RPC22981
Encompass_Preferred
Recombinant Human CD166 antigen (ALCAM), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: ALCAM. Target Synonyms: Activated leukocyte cell adhesion moleculeCD_antigen: CD166. Accession Number: Q13740. Expression Region: 28~516aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 70.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5220