Learn More
Biomatik Corporation Recombinant Human Lymphocyte antigen 6E (LY6E), 100UG
Supplier:  Biomatik Corporation RPC23009
Encompass_Preferred
Recombinant Human Lymphocyte antigen 6E (LY6E) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: LY6E. Target Synonyms: Retinoic acid-induced gene E proteinRIG-EStem cell antigen 2SCA-2Thymic shared antigen 1TSA-1. Accession Number: Q16553. Expression Region: 21~101aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 24.5kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5222