Learn More
Biomatik Corporation Recombinant Human Serotonin N-acetyltransferase (AANAT), 100UG
Supplier:  Biomatik Corporation RPC23042
Encompass_Preferred
Recombinant Human Serotonin N-acetyltransferase (AANAT) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: AANAT. Target Synonyms: Aralkylamine N-acetyltransferase. Accession Number: Q16613. Expression Region: 1~207aa. Tag Info: N-Terminal 10Xhis-Gst-Tagged. Theoretical MW: 51.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5224