Learn More
Biomatik Corporation Recombinant Anaplasma phagocytophilum 50S ribosomal protein L22 (rplV), 100UG
Supplier:  Biomatik Corporation RPC23076
Encompass_Preferred
Recombinant Anaplasma phagocytophilum 50S ribosomal protein L22 (rplV) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Anaplasma phagocytophilum (strain HZ). Target Name: rplV. Target Synonyms: rplV; APH_0285; 50S ribosomal protein L22. Accession Number: Q2GL54. Expression Region: 1~112aa. Tag Info: N-Terminal 10Xhis-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 17.2kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5226