Learn More
Biomatik Corporation Recombinant Bordetella pertussis Pertactin autotransporter (prn), partial, 100UG
Supplier:  Biomatik Corporation RPC20111
Encompass_Preferred
Recombinant Bordetella pertussis Pertactin autotransporter (prn), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Bordetella pertussis (strain Tohama I / ATCC BAA-589 / NCTC 13251). Target Name: prn. Target Synonyms: P.93. Accession Number: P14283. Expression Region: 632~910aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged. Theoretical MW: 45.8kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4599