Learn More
Biomatik Corporation Recombinant Mouse Angiogenin-4 (Ang4), 100UG
Supplier:  Biomatik Corporation RPC23089
Encompass_Preferred
Recombinant Mouse Angiogenin-4 (Ang4) is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Ang4. Target Synonyms: Ang4Angiogenin-4; EC 3.1.27. Accession Number: Q3TMQ6. Expression Region: 25~144aa. Tag Info: N-Terminal 6Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 31.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5229