Learn More
Biomatik Corporation Recombinant Mouse Putative phospholipase B-like 2 (Plbd2), 100UG
Supplier:  Biomatik Corporation RPC23093
Encompass_Preferred
Recombinant Mouse Putative phospholipase B-like 2 (Plbd2) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Mouse (Mus musculus). Target Name: Plbd2. Target Synonyms: 66.3 kDa protein76 kDa protein; p76LAMA-like protein 2Lamina ancestor homolog 2; Phospholipase B domain-containing protein 2. Accession Number: Q3TCN2. Expression Region: 47~594aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 65.9kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5230