Learn More
Biomatik Corporation Recombinant Triticum aestivum Trypsin/alpha-amylase inhibitor CMx1/CMx3, 100UG
Supplier:  Biomatik Corporation RPC23106
Encompass_Preferred
Recombinant Triticum aestivum Trypsin/alpha-amylase inhibitor CMx1/CMx3 is a purified Recombinant Protein. Purity: >85% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Triticum aestivum (Wheat). Target Name: Triticum aestivum Trypsin/alpha-amylase inhibitor CMx1/CMx3. Target Synonyms: ITRL-1/ITRL-3. Accession Number: Q43723. Expression Region: 25~121aa. Tag Info: N-Terminal 10Xhis-Sumo-Tagged And C-Terminal Myc-Tagged. Theoretical MW: 31.4kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5235